Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPY19L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172774.1 | NP_001166245.1 | 11 | 1343 | ||
NM_207325.2 | NP_997208.2 | 11 | 1369 | |||
XM_006723045.1 | XP_006723108.1 | 8 | 885 | |||
XM_011526526.2 | XP_011524828.1 | 11 | 1324 | |||
XM_017026366.1 | XP_016881855.1 | 10 | 1258 | |||
XM_017026367.1 | XP_016881856.1 | 11 | 1283 | |||
GenBank mRNA | AK126264.1 | - | 5 | 444 | ||
AK126757.1 | - | 11 | 1360 | |||
AK129497.1 | - | 11 | 1324 | |||
AL833713.1 | - | 1 | 65 | |||
BC131725.1 | - | 10 | 1191 | 40123868 | ||
CR749459.1 | - | 5 | 539 |