Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPY19L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172774.1 | NP_001166245.1 | 19 | 3346 | ||
NM_207325.2 | NP_997208.2 | 19 | 3372 | |||
XM_006723045.1 | XP_006723108.1 | 16 | 2888 | |||
XM_011526526.2 | XP_011524828.1 | 19 | 3327 | |||
XM_017026366.1 | XP_016881855.1 | 18 | 3261 | |||
XM_017026367.1 | XP_016881856.1 | 19 | 3286 | |||
GenBank mRNA | AK125846.1 | - | 3 | 1490 | ||
AK126757.1 | - | 19 | 3363 | |||
AK129497.1 | - | 19 | 3327 | |||
AL833713.1 | - | 9 | 3325 | |||
BC131725.1 | - | 18 | 3194 | 40123868 | ||
CR749459.1 | - | 13 | 2542 |