Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004701.3 | NP_004692.1 | 4 | 480 | ||
GenBank mRNA | AB020981.1 | - | 4 | 365 | ||
AF002822.1 | - | 4 | 315 | |||
AF087910.1 | - | 4 | 422 | |||
AK001404.1 | - | 4 | 444 | |||
AK222611.1 | - | 4 | 413 | |||
AL080146.1 | - | 4 | 433 | |||
AM851054.1 | - | 4 | 296 | |||
BC105086.1 | - | 4 | 428 | 8144089 | ||
BC105112.1 | - | 4 | 428 | 8144115 | ||
BT019458.1 | - | 4 | 296 | |||
CR533527.1 | - | 4 | 296 |