Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113402.1 | NP_001106873.1 | 4 | 568 | ||
NM_001278411.1 | NP_001265340.1 | 4 | 548 | |||
NM_001278412.1 | NP_001265341.1 | 6 | 777 | |||
NR_103522.1 | 5 | 644 | ||||
NR_103523.1 | 3 | 435 | ||||
XM_006719054.3 | XP_006719117.1 | 3 | 3861 | |||
XM_011520604.2 | XP_011518906.1 | 3 | 334 | |||
XM_017018963.1 | XP_016874452.1 | 6 | 617 | |||
XM_017018964.1 | XP_016874453.1 | 4 | 550 | |||
XM_017018965.1 | XP_016874454.1 | 5 | 642 | |||
GenBank mRNA | AK123033.1 | - | 4 | 415 | ||
AK302112.1 | - | 6 | 624 | |||
AK311214.1 | - | 3 | 361 | |||
AK311249.1 | - | 5 | 491 | |||
AL833434.1 | - | 4 | 418 | |||
BC037897.2 | - | 4 | 472 | 5275850 | ||
BC045831.1 | - | 4 | 512 | 5265088 | ||
BC067906.1 | - | 4 | 423 | 5314530 | ||
BI464068.1 | - | 4 | 500 | |||
DC325581.1 | - | 4 | 474 |