Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113402.1 | NP_001106873.1 | 7 | 1332 | ||
NM_001278411.1 | NP_001265340.1 | 7 | 1312 | |||
NM_001278412.1 | NP_001265341.1 | 9 | 1541 | |||
NR_103522.1 | 9 | 1495 | ||||
NR_103523.1 | 6 | 1199 | ||||
XM_006719054.3 | XP_006719117.1 | 6 | 4625 | |||
XM_011520604.2 | XP_011518906.1 | 6 | 1098 | |||
XM_017018963.1 | XP_016874452.1 | 9 | 1381 | |||
XM_017018964.1 | XP_016874453.1 | 7 | 1314 | |||
XM_017018965.1 | XP_016874454.1 | 8 | 1406 | |||
GenBank mRNA | AK094708.1 | - | 1 | 2032 | ||
AK123033.1 | - | 7 | 1179 | |||
AL833434.1 | - | 7 | 1182 | |||
BC037897.2 | - | 7 | 1236 | 5275850 | ||
BC045831.1 | - | 7 | 1276 | 5265088 | ||
BC067906.1 | - | 7 | 1187 | 5314530 |