Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278562.1 | NP_001265491.1 | 3 | 239 | ||
NM_207338.3 | NP_997221.2 | 4 | 530 | |||
NR_103739.1 | 4 | 403 | ||||
XM_017021999.1 | XP_016877488.1 | 4 | 449 | |||
GenBank mRNA | AK090598.1 | - | 3 | 224 | ||
AK311441.1 | - | 4 | 403 | |||
AY358729.1 | - | 4 | 530 |