Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004704.4 | NP_004695.1 | 2 | 202 | ||
GenBank mRNA | AJ001340.1 | - | 2 | 162 | ||
AK313696.1 | - | 2 | 202 | |||
BC001113.1 | - | 2 | 185 | 3050186 | ||
BC009879.2 | - | 2 | 167 | 3954043 | ||
BC010048.1 | - | 2 | 185 | 3832948 | ||
BC021032.1 | - | 2 | 185 | |||
BC023662.2 | - | 2 | 133 | 4898690 |