Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204118.1 | NP_001191047.1 | 10, 11 | 717 | ||
NM_207390.3 | NP_997273.3 | 10, 11 | 717 | |||
NR_109785.1 | 10, 11 | 717 | ||||
XM_017026785.1 | XP_016882274.1 | 11, 12 | 1261 | |||
XM_017026786.1 | XP_016882275.1 | 11, 12 | 1125 | |||
XM_017026787.1 | XP_016882276.1 | 12, 13 | 1480 | |||
XM_017026788.1 | XP_016882277.1 | 11, 12 | 1237 | |||
XM_017026789.1 | XP_016882278.1 | 7, 8 | 939 | |||
XM_017026790.1 | XP_016882279.1 | 10, 11 | 1213 | |||
XM_017026792.1 | XP_016882281.1 | 11, 12 | 1265 | |||
XM_017026793.1 | XP_016882282.1 | 11, 12 | 1268 | |||
GenBank mRNA | BC140848.1 | - | 10, 11 | 678 | 9021716 | |
BC144665.1 | - | 10, 11 | 678 | 9053196 |