Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTDAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244847.1 | NP_001231776.1 | 2 | 195 | ||
NM_207392.2 | NP_997275.1 | 2 | 195 | |||
GenBank mRNA | AJ293408.1 | - | 2 | 156 | ||
AY359007.1 | - | 2 | 118 | |||
BC130501.1 | - | 2 | 195 | 40146532 | ||
W95920.1 | - | 2 | 104 |