Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004577.3 | NP_004568.2 | 5 | 962 | ||
XM_005271773.1 | XP_005271830.1 | 5 | 1032 | |||
XM_005271774.1 | XP_005271831.1 | 6 | 1148 | |||
XM_005271775.1 | XP_005271832.1 | 5 | 988 | |||
XM_005271776.1 | XP_005271833.1 | 6 | 1096 | |||
XM_006715760.1 | XP_006715823.1 | 6 | 1146 | |||
XM_011515461.1 | XP_011513763.1 | 6 | 1102 | |||
XM_017012466.1 | XP_016867955.1 | 7 | 1262 | |||
XM_017012467.1 | XP_016867956.1 | 7 | 1358 | |||
GenBank mRNA | AK223508.1 | - | 2 | 252 | ||
AK315235.1 | - | 4 | 574 | |||
BC063614.1 | - | 5 | 810 | 3850060 | ||
BX537439.1 | - | 5 | 932 |