Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004578.3 | NP_004569.2 | 3, 4 | 497 | ||
NR_073545.1 | 3, 4 | 497 | ||||
GenBank mRNA | AF498934.1 | - | 3, 4 | 219 | ||
AK223009.1 | - | 3, 4 | 262 | |||
AK313807.1 | - | 3, 4 | 470 | |||
AY585832.1 | - | 3, 4 | 426 | |||
BC002438.1 | - | 3, 4 | 290 | 3346455 | ||
BC004309.1 | - | 3, 4 | 427 | 3627110 | ||
BI828974.1 | - | 3, 4 | 381 | |||
HY142709.1 | - | 3 | 477 | |||
KC407675.1 | - | 3, 4 | 219 | |||
M28211.1 | - | 3, 4 | 274 | |||
X13902.1 | - | 3, 4 | 330 |