Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207426.2 | NP_997309.2 | 2 | 2995 | ||
GenBank mRNA | AK128865.1 | - | 2 | 2595 |