Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207437.3 | NP_997320.2 | 8 | 927 | ||
XM_005253555.3 | XP_005253612.1 | 8 | 1419 | |||
XM_011538014.2 | XP_011536316.1 | 8 | 1419 | |||
XM_011538015.2 | XP_011536317.1 | 8 | 1419 | |||
XM_011538016.2 | XP_011536318.1 | 8 | 1419 | |||
XM_011538017.2 | XP_011536319.1 | 8 | 1419 | |||
XM_011538020.2 | XP_011536322.1 | 8 | 1427 | |||
XM_017018960.1 | XP_016874449.1 | 8 | 1419 | |||
XM_017018961.1 | XP_016874450.1 | 8 | 1419 | |||
GenBank mRNA | AK125475.1 | - | 3 | 441 | ||
BC144575.1 | - | 3 | 417 | |||
BC150622.1 | - | 3 | 417 |