Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004593.2 | NP_004584.1 | 2 | 436 | ||
XM_005247703.1 | XP_005247760.1 | 3 | 604 | |||
XM_011513072.2 | XP_011511374.1 | 3 | 3406 | |||
XM_017007033.1 | XP_016862522.1 | 2 | 1582 | |||
GenBank mRNA | AB209208.1 | - | 3 | 3393 | ||
AK315632.1 | - | 2 | 420 | |||
BC000160.2 | - | 2 | 303 | 2984771 | ||
BC000451.2 | - | 2 | 302 | 2821443 | ||
BC005898.1 | - | 2 | 287 | 4046349 | ||
CR749214.1 | - | 1 | 2463 | |||
U61267.1 | - | 2 | 281 | |||
U68063.1 | - | 2 | 289 |