Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004616.2 | NP_004607.1 | 6 | 604 | ||
XM_006719583.3 | XP_006719646.1 | 6 | 558 | |||
XM_017019913.1 | XP_016875402.1 | 5 | 640 | |||
GenBank mRNA | AK223022.1 | - | 5 | 532 | ||
AK313112.1 | - | 5 | 587 | |||
BC005246.1 | - | 6 | 604 | 3934330 | ||
BC070168.1 | - | 10 | 978 | |||
M35252.1 | - | 5 | 562 |