Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC401554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135124.1 | 2 | 2087 | |||
GenBank mRNA | AK128673.1 | - | 2 | 2087 | ||
BC132936.1 | - | 1 | 799 |