Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145313.2 | NP_001138785.1 | 9, 10 | 1090 | ||
NM_001287191.1 | NP_001274120.1 | 8, 9 | 994 | |||
NM_001287192.1 | NP_001274121.1 | 8, 9 | 994 | |||
XM_005252254.2 | XP_005252311.1 | 9, 10 | 1323 | |||
XM_005252257.3 | XP_005252314.1 | 8, 9 | 989 | |||
XM_005252260.3 | XP_005252317.1 | 7, 8 | 956 | |||
XM_011519077.2 | XP_011517379.1 | 10, 11 | 981 | |||
XM_011519078.2 | XP_011517380.2 | 9, 10 | 988 | |||
XM_011519079.2 | XP_011517381.2 | 9, 10 | 990 | |||
XM_011519080.2 | XP_011517382.1 | 3, 4 | 1686 | |||
XM_011519081.2 | XP_011517383.1 | 3, 4 | 1671 | |||
XM_017015182.1 | XP_016870671.1 | 9, 10 | 1323 | |||
XM_017015183.1 | XP_016870672.1 | 8, 9 | 989 | |||
XM_017015184.1 | XP_016870673.1 | 4, 5 | 499 | |||
XM_017015185.1 | XP_016870674.1 | 9, 10 | 996 | |||
XM_017015186.1 | XP_016870675.1 | 3, 4 | 339 | |||
XM_017015187.1 | XP_016870676.1 | 8, 9 | 989 | |||
GenBank mRNA | AK296433.1 | - | 8, 9 | 905 | ||
AK299350.1 | - | 9, 10 | 1022 | |||
AK299491.1 | - | 8, 9 | 905 | |||
AK309817.1 | - | 9, 10 | 1331 | |||
BC036746.1 | - | 3, 4 | 433 | 4384472 |