Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291019.1 | NP_001277948.1 | 3 | 1096 | ||
NM_001291020.1 | NP_001277949.1 | 4 | 898 | |||
NM_007547.4 | NP_031573.2 | 3 | 1096 | |||
GenBank mRNA | AB018194.1 | - | 3 | 662 | ||
AF072543.1 | - | 3 | 652 | |||
AF332079.1 | - | 3 | 662 | |||
AF332080.1 | - | 3 | 662 | |||
AK159617.1 | - | 4 | 903 | |||
AK193777.1 | - | 1 | 118 | |||
AK197651.1 | - | 1 | 118 | |||
AK201200.1 | - | 1 | 118 | |||
BC062197.1 | - | 3 | 687 | 5368250 | ||
D85785.1 | - | 3 | 946 | |||
D87967.1 | - | 3 | 1061 | |||
D87968.1 | - | 3 | 1061 | |||
U89694.1 | - | 3 | 673 | |||
Y10349.1 | - | 3 | 707 |