Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008831.4 | NP_032857.1 | 4 | 427 | ||
GenBank mRNA | AK002714.1 | - | 3 | 373 | ||
AK010619.1 | - | 4 | 370 | |||
AK150956.1 | - | 4 | 372 | |||
AK151073.1 | - | 4 | 372 | |||
AK152933.1 | - | 4 | 374 | |||
AK160490.1 | - | 4 | 366 | |||
AK168248.1 | - | 4 | 375 | |||
AK168656.1 | - | 4 | 366 | |||
AK190324.1 | - | 1 | 54 | |||
BC083354.1 | - | 3 | 349 | 5715690 | ||
BC089034.1 | - | 3 | 351 | 30675988 | ||
X78682.1 | - | 3 | 355 |