Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pick1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045558.1 | NP_001039023.1 | 4 | 505 | ||
NM_008837.3 | NP_032863.2 | 4 | 564 | |||
GenBank mRNA | AK050224.1 | - | 4 | 476 | ||
AK090155.1 | - | 497 | ||||
BC023088.1 | - | 4 | 471 | |||
BC048788.1 | - | 4 | 450 | 6488048 | ||
BU057774.1 | - | 4 | 467 | |||
CA468666.1 | - | 4 | 385 | |||
CK791908.1 | - | 4 | 505 | |||
Z46720.1 | - | 3 | 253 |