Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3cd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029837.2 | NP_001025008.2 | 8 | 1249 | ||
NM_001164049.1 | NP_001157521.1 | 8 | 1276 | |||
NM_001164050.1 | NP_001157522.1 | 8 | 1276 | |||
NM_001164051.1 | NP_001157523.1 | 8 | 1300 | |||
NM_001164052.1 | NP_001157524.1 | 6 | 1003 | |||
NM_008840.3 | NP_032866.2 | 9 | 1451 | |||
GenBank mRNA | AK040867.1 | - | 8 | 1211 | ||
AK149935.1 | - | 9 | 1350 | |||
AK150649.1 | - | 8 | 1177 | |||
AK151482.1 | - | 8 | 1177 | |||
AK152661.1 | - | 8 | 1177 | |||
AK153304.1 | - | 8 | 1211 | |||
AK155835.1 | - | 8 | 1162 | |||
AK171025.1 | - | 8 | 1259 | |||
AK172222.1 | - | 8 | 1153 | |||
BC035203.1 | - | 8 | 1266 | 4192906 | ||
CT010349.1 | - | 6 | 971 | |||
U86587.1 | - | 6 | 971 |