Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004626.2 | NP_004617.2 | 4, 5 | 1021 | ||
XM_005274231.1 | XP_005274288.1 | 5, 6 | 1277 | |||
XM_011545238.1 | XP_011543540.1 | 5, 6 | 1307 | |||
XM_011545239.2 | XP_011543541.1 | 5, 6 | 1515 | |||
XM_011545240.2 | XP_011543542.1 | 6, 7 | 1471 | |||
XM_011545241.2 | XP_011543543.1 | 4, 5 | 759 | |||
XM_017018241.1 | XP_016873730.1 | 3, 4 | 1779 | |||
XM_017018242.1 | XP_016873731.1 | 3, 4 | 1614 | |||
XM_017018243.1 | XP_016873732.1 | 2, 3 | 433 | |||
GenBank mRNA | AB070218.1 | - | 4, 5 | 960 | ||
AK313570.1 | - | 4, 5 | 1023 | |||
BC074790.2 | - | 4, 5 | 940 | 30915564 | ||
BC074791.2 | - | 4, 5 | 940 | 30915347 | ||
BC113386.1 | - | 4, 5 | 941 | 8322438 | ||
BC113388.1 | - | 4, 5 | 941 | 8322440 | ||
BT019492.1 | - | 4, 5 | 898 | |||
JF800675.1 | - | 4, 5 | 622 | |||
JQ715612.1 | - | 4, 5 | 777 | |||
JQ715613.1 | - | 4, 5 | 586 |