Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008841.2 | NP_032867.2 | 8 | 1381 | ||
GenBank mRNA | AK146195.1 | - | 8 | 1371 | ||
AK148122.1 | - | 8 | 1378 | |||
AK216667.1 | - | 3 | 316 | |||
BC006796.1 | - | 8 | 1372 | 2581969 | ||
BC085501.1 | - | 7 | 1304 | 6816009 | ||
Y13569.1 | - | 8 | 1350 |