Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004628.4 | NP_004619.3 | 2 | 272 | ||
NR_027299.1 | 2 | 272 | ||||
XM_011534093.2 | XP_011532395.1 | 2 | 229 | |||
GenBank mRNA | AK289761.1 | - | 2 | 201 | ||
AK295711.1 | - | 2 | 201 | |||
AK311039.1 | - | 2 | 201 | |||
AL710884.1 | - | 2 | 239 | |||
BC016620.1 | - | 2 | 183 | 4509957 | ||
D21089.1 | - | 2 | 273 | |||
HM113488.1 | - | 2 | 172 | |||
KJ534962.1 | - | 2 | 168 | |||
KJ535085.1 | - | 2 | 168 |