Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pja1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083110.2 | NP_001076579.1 | 2 | 1971 | ||
NM_001290555.1 | NP_001277484.1 | 2 | 2257 | |||
NM_001290556.1 | NP_001277485.1 | 3 | 1431 | |||
NM_008853.3 | NP_032879.2 | 3 | 1422 | |||
GenBank mRNA | AF335250.2 | - | 2 | 1940 | ||
AF335251.2 | - | 3 | 1295 | |||
AF335252.1 | - | 3 | 684 | |||
AK005373.1 | - | 2 | 1878 | |||
AK135382.1 | - | 2 | 1956 | |||
AK145943.1 | - | 2 | 2214 | |||
BC008148.1 | - | 1 | 1245 | |||
BC025975.1 | - | 2 | 1919 | 3983750 | ||
BC037616.1 | - | 2 | 1894 | 5401000 |