Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008872.2 | NP_032898.2 | 10 | 1028 | ||
GenBank mRNA | AK135857.1 | - | 10 | 1019 | ||
AK135965.1 | - | 10 | 1020 | |||
AK141665.1 | - | 10 | 1017 | |||
AK159620.1 | - | 10 | 1020 | |||
AK166639.1 | - | 10 | 1018 | |||
BC011256.1 | - | 10 | 1025 | 4038404 | ||
BC057967.1 | - | 10 | 1008 | 4979664 | ||
BC061508.1 | - | 10 | 1009 | 5352018 | ||
J03520.1 | - | 10 | 1011 |