Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004643.3 | NP_004634.1 | 2 | 1701 | ||
GenBank mRNA | AK293633.1 | - | 1 | 80 | ||
BC010939.1 | - | 2 | 427 | 4333442 | ||
BI830835.1 | - | 2 | 602 | |||
BX247976.1 | - | 2 | 425 |
Gene Symbol: BCL2L2-PABPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199864.1 | NP_001186793.1 | 4 | 729 | ||
GenBank mRNA | BC011637.1 | - | 4 | 665 |