Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008881.2 | NP_032907.1 | 2 | 489 | ||
GenBank mRNA | BC138023.1 | - | 1 | 281 | 8861042 | |
D86948.1 | - | 2 | 473 |