Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA795914.1 | - | 1 | 152 | ||
AK076005.1 | - | 5 | 1628 | |||
AK171745.1 | - | 5 | 1625 | |||
BC010765.1 | - | 5 | 1651 | 4225316 | ||
M32240.1 | - | 5 | 1678 |