Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008886.2 | NP_032912.2 | 2 | 171 | ||
GenBank mRNA | AK014190.1 | - | 2 | 149 | ||
AK017847.1 | - | 2 | 147 | |||
AK034866.1 | - | 2 | 147 | |||
AK083049.1 | - | 2 | 128 | |||
AK146365.1 | - | 2 | 184 | |||
BC058189.1 | - | 2 | 179 | |||
BC066110.1 | - | 2 | 171 | |||
BC083502.1 | - | 2 | 131 | |||
BC086637.1 | - | 2 | 147 | |||
BC110684.1 | - | 2 | 124 | |||
BC139774.1 | - | 2 | 177 | |||
BC141287.1 | - | 2 | 147 | 9055899 |