Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008886.2 | NP_032912.2 | 6 | 622 | ||
GenBank mRNA | AK017847.1 | - | 6 | 598 | ||
AK034866.1 | - | 6 | 598 | |||
AK083049.1 | - | 6 | 579 | |||
AK146365.1 | - | 6 | 635 | |||
BC022179.1 | - | 6 | 590 | |||
BC066110.1 | - | 6 | 622 | |||
BC083502.1 | - | 6 | 582 | |||
BC086637.1 | - | 6 | 598 | |||
BC110684.1 | - | 6 | 575 | |||
BC139774.1 | - | 6 | 628 | |||
BC141287.1 | - | 6 | 598 | 9055899 |