Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008886.2 | NP_032912.2 | 10, 11 | 1208 | ||
GenBank mRNA | AK017847.1 | - | 10, 11 | 1184 | ||
AK034866.1 | - | 10, 11 | 1184 | |||
AK083049.1 | - | 10, 11 | 1165 | |||
AK146365.1 | - | 10, 11 | 1221 | |||
BC022179.1 | - | 10, 11 | 1176 | |||
BC058189.1 | - | 10, 11 | 1216 | |||
BC066110.1 | - | 10, 11 | 1208 | |||
BC083502.1 | - | 10, 11 | 1168 | |||
BC086637.1 | - | 10, 11 | 1184 | |||
BC110684.1 | - | 10, 11 | 1161 | |||
BC139774.1 | - | 10, 11 | 1214 | |||
BC141287.1 | - | 10, 11 | 1184 | 9055899 |