Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008909.2 | NP_032935.2 | 22 | 3919 | ||
GenBank mRNA | AF126834.1 | - | 22 | 3859 | ||
AK028144.1 | - | 1 | 49 |