Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP9X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039590.2 | NP_001034679.2 | 6, 7 | 1295 | ||
NM_001039591.2 | NP_001034680.2 | 6, 7 | 1295 | |||
XM_005272675.4 | XP_005272732.1 | 6, 7 | 1510 | |||
XM_005272676.4 | XP_005272733.1 | 6, 7 | 1464 | |||
GenBank mRNA | AK294828.1 | - | 5, 6 | 696 | ||
X98296.1 | - | 5, 6 | 700 |