Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008917.3 | NP_032943.2 | 9 | 1496 | ||
GenBank mRNA | AF071025.1 | - | 9 | 1512 | ||
AF087568.1 | - | 9 | 1476 | |||
AK014561.1 | - | 9 | 1478 | |||
AK151055.1 | - | 9 | 1471 | |||
AK151107.1 | - | 9 | 1482 | |||
AK152940.1 | - | 9 | 1482 | |||
AK153106.1 | - | 9 | 1477 | |||
AK153316.1 | - | 9 | 1478 | |||
AK169579.1 | - | 10 | 1571 | |||
AK169658.1 | - | 10 | 1576 | |||
AK171668.1 | - | 9 | 1477 | |||
BC099945.1 | - | 1 | 2379 |