Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293650.1 | NP_001280579.1 | 14, 15 | 1425 | ||
NM_001293651.1 | NP_001280580.1 | 14, 15 | 1408 | |||
NM_008925.2 | NP_032951.1 | 14, 15 | 1404 | |||
GenBank mRNA | AK137661.1 | - | 14, 15 | 1371 | ||
AK153931.1 | - | 14, 15 | 1379 | |||
AK159705.1 | - | 14, 15 | 1375 | |||
BC009816.1 | - | 14, 15 | 1358 | 4008182 | ||
CX207653.1 | - | 6, 7 | 544 | |||
U92794.1 | - | 14, 15 | 1357 |