Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164058.1 | NP_001157530.1 | 6 | 513 | ||
NM_008930.2 | NP_032956.1 | 6 | 510 | |||
GenBank mRNA | AF011381.1 | - | 6 | 502 | ||
AF020525.1 | - | 6 | 484 | |||
AK028371.1 | - | 6 | 511 | |||
AK141309.1 | - | 6 | 514 | |||
BB611867.1 | - | 6 | 511 | |||
BC051671.1 | - | 6 | 495 | 30018275 |