Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008937.2 | NP_032963.1 | 2 | 576 | ||
GenBank mRNA | AF061576.1 | - | 1 | 163 | ||
AK052889.1 | - | 2 | 610 | |||
AK143179.1 | - | 2 | 400 | |||
BC051411.1 | - | 2 | 576 | 6490801 |