Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008938.1 | NP_032964.1 | 3 | 1789 | ||
GenBank mRNA | BC026484.1 | - | 1 | 2122 | ||
X14770.1 | - | 3 | 1789 |