Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008943.2 | NP_032969.1 | 4 | 715 | ||
GenBank mRNA | AF149111.1 | - | 2 | 227 | ||
AK134232.1 | - | 4 | 398 | |||
AK134615.1 | - | 4 | 437 | |||
AK148101.1 | - | 4 | 436 | |||
AK165984.1 | - | 4 | 436 | |||
BC014744.1 | - | 4 | 466 | 4511140 | ||
BC030409.1 | - | 4 | 390 | 5133302 | ||
BC071233.1 | - | 4 | 454 | 3673609 | ||
L42177.1 | - | 4 | 411 |