Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008943.2 | NP_032969.1 | 12 | 2778 | ||
GenBank mRNA | AK148101.1 | - | 12 | 2499 | ||
AK169963.1 | - | 8 | 1905 | |||
AK205585.1 | - | 1 | 164 | |||
BC071233.1 | - | 12 | 2517 | 3673609 |