Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008948.2 | NP_032974.2 | 12 | 1600 | ||
GenBank mRNA | AK002542.1 | - | 12 | 1321 | ||
AK151370.1 | - | 12 | 1320 | |||
AK152018.1 | - | 12 | 1339 | |||
AK167002.1 | - | 12 | 1548 | |||
AK167061.1 | - | 12 | 1545 | |||
AK168263.1 | - | 11 | 1269 | |||
AK168616.1 | - | 12 | 1320 | |||
BC005783.1 | - | 12 | 1489 | 3603053 | ||
D49686.1 | - | 12 | 1544 |