Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008950.1 | NP_032976.1 | 7 | 761 | ||
GenBank mRNA | AK010505.1 | - | 7 | 781 | ||
AK012294.1 | - | 7 | 794 | |||
AK135451.1 | - | 7 | 771 | |||
AK145703.1 | - | 7 | 765 | |||
BC004052.1 | - | 3 | 456 | |||
BC030840.1 | - | 7 | 793 | 3994523 | ||
Z54219.1 | - | 7 | 761 |