Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008963.2 | NP_032989.2 | 1 | 178 | ||
GenBank mRNA | AB006361.1 | - | 1 | 160 | ||
AK131859.1 | - | 1 | 178 | |||
BC043015.1 | - | 1 | 164 | 5683706 | ||
X89222.1 | - | 1 | 149 |