Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008968.3 | NP_032994.1 | 8, 9 | 1334 | ||
GenBank mRNA | AB001607.1 | - | 8, 9 | 1252 | ||
AK048059.1 | - | 1387 | ||||
AK076585.1 | - | 8, 9 | 1335 | |||
BC062151.1 | - | 8, 9 | 1341 | 6512164 |