Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgis | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008968.3 | NP_032994.1 | 9, 10 | 1477 | ||
GenBank mRNA | AB001607.1 | - | 9, 10 | 1395 | ||
AK076585.1 | - | 9, 10 | 1478 | |||
BC062151.1 | - | 9, 10 | 1484 | 6512164 |