Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008973.2 | NP_032999.1 | 5 | 2009 | ||
GenBank mRNA | AK011346.1 | - | 5 | 866 | ||
AK161964.1 | - | 5 | 700 | |||
BC002064.1 | - | 5 | 578 | 3488877 | ||
BC061695.1 | - | 5 | 809 | 6313227 | ||
D90225.1 | - | 5 | 753 | |||
EF437379.1 | - | 2 | 240 |