Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166388.1 | NP_001159860.1 | 6 | 2457 | ||
NM_001166389.1 | NP_001159861.1 | 6 | 2188 | |||
NM_001166390.1 | NP_001159862.1 | 5 | 1289 | |||
NM_008975.3 | NP_033001.2 | 6 | 2257 | |||
GenBank mRNA | AK014601.1 | - | 6 | 1871 |