Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igdcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008988.2 | NP_033014.1 | 3, 4 | 824 | ||
GenBank mRNA | AF026465.1 | - | 3, 4 | 824 | ||
AK051027.1 | - | 3, 4 | 825 | |||
BC053057.1 | - | 3, 4 | 801 | 5698028 | ||
BC086481.1 | - | 3, 4 | 808 | 6505546 |