Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008992.2 | NP_033018.2 | 4, 5 | 565 | ||
GenBank mRNA | AJ001166.1 | - | 4, 5 | 469 | ||
AK028420.1 | - | 4, 5 | 470 | |||
BC050102.1 | - | 4, 5 | 673 | 30012556 |